Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5,6-dimethyl-1,2,4-triazine 98.0+%, TCI America™

Click to view available options
Quantity:
25g
17584-12-2
C5H8N4
124.147
MFCD00006460
UIKGLXJNZXSPGV-UHFFFAOYSA-N
3-amino-5,6-dimethyl-1,2,4-triazine,1,2,4-triazin-3-amine, 5,6-dimethyl,dimethyl-1,2,4-triazin-3-amine,3-amino-5,6-dimethyl-as-triazine,unii-y36asr52nu,as-triazine, 3-amino-5,6-dimethyl,y36asr52nu,5,6-dimethyl-1,2,4-triazin-3-ylamine,5,6-dimethyl-1,2,4-triazine-3-ylamine,acmc-1bolj
87163
5,6-dimethyl-1,2,4-triazin-3-amine
CC1=C(N=NC(=N1)N)C
17584-12-2
C5H8N4
124.147
MFCD00006460
UIKGLXJNZXSPGV-UHFFFAOYSA-N
3-amino-5,6-dimethyl-1,2,4-triazine,1,2,4-triazin-3-amine, 5,6-dimethyl,dimethyl-1,2,4-triazin-3-amine,3-amino-5,6-dimethyl-as-triazine,unii-y36asr52nu,as-triazine, 3-amino-5,6-dimethyl,y36asr52nu,5,6-dimethyl-1,2,4-triazin-3-ylamine,5,6-dimethyl-1,2,4-triazine-3-ylamine,acmc-1bolj
87163
5,6-dimethyl-1,2,4-triazin-3-amine
CC1=C(N=NC(=N1)N)C
Melting Point 210°C
Color Undesignated
Quantity 25g
Formula Weight 124.15
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 3-Amino-5,6-dimethyl-1,2,4-triazine

RTECSNumber : XY3177100

TSCA : No