Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-bromo-1H-indazole, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
61272-71-7
C7H6BrN3
212.05
MFCD03426696
OMPYFDJVSAMSMA-UHFFFAOYSA-N
3-amino-5-bromo-1h-indazole,3-amino-5-bromoindazole,5-bromo-1h-indazol-3-ylamine,1h-indazol-3-amine, 5-bromo,5-bromo-1h-indazole-3-ylamine,pubchem18573,3-amino-5-bromo-indazole,acmc-1b3h9,5-bromo-1h-indazole-3-amine
817910
5-bromo-1H-indazol-3-amine
C1=CC2=C(C=C1Br)C(=NN2)N
61272-71-7
C7H6BrN3
212.05
MFCD03426696
OMPYFDJVSAMSMA-UHFFFAOYSA-N
3-amino-5-bromo-1h-indazole,3-amino-5-bromoindazole,5-bromo-1h-indazol-3-ylamine,1h-indazol-3-amine, 5-bromo,5-bromo-1h-indazole-3-ylamine,pubchem18573,3-amino-5-bromo-indazole,acmc-1b3h9,5-bromo-1h-indazole-3-amine
817910
5-bromo-1H-indazol-3-amine
C1=CC2=C(C=C1Br)C(=NN2)N
Melting Point 237°C to 238°C
Packaging Glass Bottle
Color Colorless
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 212.05
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 3-Amino-5-bromo-1H-indazole
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning