Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-5-bromopyridine, 98%

Click to view available options
Quantity:
1g
5g
25g
13535-01-8
C5H5BrN2
173.013
MFCD01646060
MDQXGHBCDCOOSM-UHFFFAOYSA-N
3-amino-5-bromopyridine,5-bromo-3-pyridinamine,5-bromo-pyridin-3-ylamine,5-bromo-3-aminopyridine,3-pyridinamine, 5-bromo,5-brompyridin-3-amin,5-bromo-3-pyridylamine,5-bromopyridin-3-ylamine,3-amino-5-bromo pyridine,pubchem1213
817681
5-bromopyridin-3-amine
C1=C(C=NC=C1Br)N
13535-01-8
C5H5BrN2
173.013
MFCD01646060
MDQXGHBCDCOOSM-UHFFFAOYSA-N
3-amino-5-bromopyridine,5-bromo-3-pyridinamine,5-bromo-pyridin-3-ylamine,5-bromo-3-aminopyridine,3-pyridinamine, 5-bromo,5-brompyridin-3-amin,5-bromo-3-pyridylamine,5-bromopyridin-3-ylamine,3-amino-5-bromo pyridine,pubchem1213
817681
5-bromopyridin-3-amine
C1=C(C=NC=C1Br)N
Quantity 1g
Melting Point 65°C to 69°C
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 3-Amino-5-bromopyridine

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures