Alfa Aesar™ 3-Amino-5-cyclobutyl-1H-pyrazole, 97%

Manufacturer: Alfa Aesar™ H50880MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50880MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 326827-21-8
    Molecular Formula C7H11N3
    Formula Weight 137.19
    MDL Number MFCD08061045
    Synonym 5-Cyclobutyl-1H-pyrazol-3-ylamine
    Chemical Name or Material 3-Amino-5-cyclobutyl-1H-pyrazole

    Specifications

    Formula Weight 137.19
    Quantity 250mg
    Molecular Formula C7H11N3
    MDL Number MFCD08061045
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    CAS 326827-21-8
    Synonym 5-Cyclobutyl-1H-pyrazol-3-ylamine
    Chemical Name or Material 3-Amino-5-cyclobutyl-1H-pyrazole
    Assay 97%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Slightly soluble in water.

    Notes
    Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit