Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-ethyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7077310
Click to view available options
Quantity:
1 g
5 g
1904-24-1
C5H9N3
111.15
MFCD08060982
AXDGPQLEVYSXNL-UHFFFAOYSA-N
3-ethyl-1h-pyrazol-5-amine, 5-amino-3-ethyl-1h-pyrazole, 3-amino-5-ethylpyrazole, 3-amino-5-ethyl-1h-pyrazole, 5-ethyl-2h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 5-ethyl, 5-ethyl-2h-pyrazol-3-amine, ethyl-5-aminopyrazole, 3-amino-5-ethypyrazole, 1h-pyrazol-5-amine, 3-ethyl
21991095
5-ethyl-1H-pyrazol-3-amine
CCC1=CC(N)=NN1
1904-24-1
C5H9N3
111.15
MFCD08060982
AXDGPQLEVYSXNL-UHFFFAOYSA-N
3-ethyl-1h-pyrazol-5-amine, 5-amino-3-ethyl-1h-pyrazole, 3-amino-5-ethylpyrazole, 3-amino-5-ethyl-1h-pyrazole, 5-ethyl-2h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 5-ethyl, 5-ethyl-2h-pyrazol-3-amine, ethyl-5-aminopyrazole, 3-amino-5-ethypyrazole, 1h-pyrazol-5-amine, 3-ethyl
21991095
5-ethyl-1H-pyrazol-3-amine
CCC1=CC(N)=NN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material 3-Amino-5-ethyl-1H-pyrazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minu

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit