Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-5-fluorobenzoic acid, 95%

Click to view available options
Quantity:
250mg
1g
5g
786616-54-4
C7H6FNO2
155.128
MFCD07779526
RYLBYCHERDTVAY-UHFFFAOYSA-N
benzoic acid, 3-amino-5-fluoro-9ci,3-amino-5-fluoro-benzoic acid,5-amino-3-fluorobenzoic acid,3-fluoro-5-aminobenzoic acid,benzoic acid,3-amino-5-fluoro,benzoic acid, 3-amino-5-fluoro
17915148
3-amino-5-fluorobenzoic acid
C1=C(C=C(C=C1N)F)C(=O)O
786616-54-4
C7H6FNO2
155.128
MFCD07779526
RYLBYCHERDTVAY-UHFFFAOYSA-N
benzoic acid, 3-amino-5-fluoro-9ci,3-amino-5-fluoro-benzoic acid,5-amino-3-fluorobenzoic acid,3-fluoro-5-aminobenzoic acid,benzoic acid,3-amino-5-fluoro,benzoic acid, 3-amino-5-fluoro
17915148
3-amino-5-fluorobenzoic acid
C1=C(C=C(C=C1N)F)C(=O)O
Quantity 1g
Formula Weight 155.13
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-Amino-5-fluorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C