Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-methylpyrazole 96.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
31230-17-8
C4H7N3
97.121
MFCD00075180
FYTLHYRDGXRYEY-UHFFFAOYSA-N
3-amino-5-methylpyrazole,3-methyl-1h-pyrazol-5-amine,1h-pyrazol-3-amine, 5-methyl,5-amino-3-methylpyrazole,3-amino-5-methyl-1h-pyrazole,5-amino-3-methyl-1h-pyrazole,5-methyl-2h-pyrazol-3-ylamine,5-methyl-2h-pyrazol-3-amine,5-methyl-1h-pyrazol-3-ylamine,5-methylpyrazol-3-amine
93146
5-methyl-1H-pyrazol-3-amine
CC1=CC(=NN1)N
31230-17-8
C4H7N3
97.121
MFCD00075180
FYTLHYRDGXRYEY-UHFFFAOYSA-N
3-amino-5-methylpyrazole,3-methyl-1h-pyrazol-5-amine,1h-pyrazol-3-amine, 5-methyl,5-amino-3-methylpyrazole,3-amino-5-methyl-1h-pyrazole,5-amino-3-methyl-1h-pyrazole,5-methyl-2h-pyrazol-3-ylamine,5-methyl-2h-pyrazol-3-amine,5-methyl-1h-pyrazol-3-ylamine,5-methylpyrazol-3-amine
93146
5-methyl-1H-pyrazol-3-amine
CC1=CC(=NN1)N
Quantity 250g
Color Undesignated
Formula Weight 97.12
Physical Form Crystal at 20°C
Percent Purity ≥96.0% (GC)
Chemical Name or Material 3-Amino-5-methylpyrazole

TSCA : No

Recommended Storage : Refrigerator