Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-methylthio-1H-1,2,4-triazole, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
45534-08-5
C3H6N4S
130.169
MFCD00005232
XGWWZKBCQLBJNH-UHFFFAOYSA-N
3-amino-5-methylthio-1h-1,2,4-triazole,1h-1,2,4-triazol-3-amine, 5-methylthio,5-methylthio-1h-1,2,4-triazol-3-amine,3-amino-5-methylthio-1,2,4-triazole,5-methylsulfanyl-4h-1,2,4-triazol-3-amine,3-methylthio-1h-1,2,4-triazol-5-amine,3-methylsulfanyl-1h-1,2,4-triazol-5-amine,5-methylsulfanyl-1h-1,2,4-triazol-3-amine,5-amino-3-methylthio-1h-1,2,4-triazole,5-methylthio-4h-1,2,4-triazol-3-amine
78471
3-methylsulfanyl-1H-1,2,4-triazol-5-amine
CSC1=NNC(=N1)N
45534-08-5
C3H6N4S
130.169
MFCD00005232
XGWWZKBCQLBJNH-UHFFFAOYSA-N
3-amino-5-methylthio-1h-1,2,4-triazole,1h-1,2,4-triazol-3-amine, 5-methylthio,5-methylthio-1h-1,2,4-triazol-3-amine,3-amino-5-methylthio-1,2,4-triazole,5-methylsulfanyl-4h-1,2,4-triazol-3-amine,3-methylthio-1h-1,2,4-triazol-5-amine,3-methylsulfanyl-1h-1,2,4-triazol-5-amine,5-methylsulfanyl-1h-1,2,4-triazol-3-amine,5-amino-3-methylthio-1h-1,2,4-triazole,5-methylthio-4h-1,2,4-triazol-3-amine
78471
3-methylsulfanyl-1H-1,2,4-triazol-5-amine
CSC1=NNC(=N1)N
Packaging Glass bottle
Color White to Beige
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 130.17
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Amino-5-methylthio-1H-1, 2, 4-triazole, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

CANCER SUSPECTED AGENT