Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-(trifluoromethyl)-1H-indazole, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
2250-53-5
C8H6F3N3
201.15
MFCD01719740
ZXZTZRVCWTWKCH-UHFFFAOYSA-N
5-trifluoromethyl-1h-indazol-3-amine,3-amino-5-trifluoromethyl-1h-indazole,1h-indazol-3-amine, 5-trifluoromethyl,1h-indazol-3-amine, 5-trifluoromethyl-9ci,1h-indazole, 3-amino-5-trifluoromethyl,5-trifluoromethyl-1h-indazole-3-ylamine,3-amino-5-trifluoromethylindazole,3-amino-5trifluoromethyl-indazole,5-trifluoromethyl-1h-indazol-3-ylamine
16736
5-(trifluoromethyl)-1H-indazol-3-amine
C1=CC2=C(C=C1C(F)(F)F)C(=NN2)N
2250-53-5
C8H6F3N3
201.15
MFCD01719740
ZXZTZRVCWTWKCH-UHFFFAOYSA-N
5-trifluoromethyl-1h-indazol-3-amine,3-amino-5-trifluoromethyl-1h-indazole,1h-indazol-3-amine, 5-trifluoromethyl,1h-indazol-3-amine, 5-trifluoromethyl-9ci,1h-indazole, 3-amino-5-trifluoromethyl,5-trifluoromethyl-1h-indazole-3-ylamine,3-amino-5-trifluoromethylindazole,3-amino-5trifluoromethyl-indazole,5-trifluoromethyl-1h-indazol-3-ylamine
16736
5-(trifluoromethyl)-1H-indazol-3-amine
C1=CC2=C(C=C1C(F)(F)F)C(=NN2)N
Melting Point 134.5°C to 135.5°C
Packaging Glass Bottle
Color White to Yellow
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 201.15
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 3-Amino-5-(trifluoromethyl)-1H-indazole
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning