Alfa Aesar™ 3-Amino-5-(trifluoromethyl)-1H-indazole, 97%

Manufacturer: Alfa Aesar™ H3299503

Quantity 1g

 View more versions of this product

Catalog No. AAH3299503

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 2250-53-5
    Molecular Formula C8H6F3N3
    Formula Weight 201.15
    MDL Number MFCD01719740
    UN Number UN2811
    Synonym 5-(Trifluoromethyl)-1H-indazol-3-amine
    Chemical Name or Material 3-Amino-5-(trifluoromethyl)-1H-indazole

    Specifications

    Formula Weight 201.15
    Molecular Formula C8H6F3N3
    Melting Point 133° to 136°C
    MDL Number MFCD01719740
    CAS 2250-53-5
    UN Number UN2811
    Quantity 1g
    Odor Odorless
    Synonym 5-(Trifluoromethyl)-1H-indazol-3-amine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number NK7772500
    Chemical Name or Material 3-Amino-5-(trifluoromethyl)-1H-indazole
    Assay 97%

    Description

    Applications
    Used as pharmaceutical intermediate.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit