Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-6-cyano-2-methylpyridine, 95%

Click to view available options
Quantity:
250mg
1g
1079054-78-6
C7H7N3
133.154
MFCD08062828
SLBIKHREAIGBFI-UHFFFAOYSA-N
5-amino-6-methylpicolinonitrile,6-cyano-2-methyl-3-aminopyridine,5-amino-6-methyl-2-pyridinecarbonitrile,5-amino-6-methyl-pyridine-2-carbonitrile
55252984
5-amino-6-methylpyridine-2-carbonitrile
CC1=C(C=CC(=N1)C#N)N
1079054-78-6
C7H7N3
133.154
MFCD08062828
SLBIKHREAIGBFI-UHFFFAOYSA-N
5-amino-6-methylpicolinonitrile,6-cyano-2-methyl-3-aminopyridine,5-amino-6-methyl-2-pyridinecarbonitrile,5-amino-6-methyl-pyridine-2-carbonitrile
55252984
5-amino-6-methylpyridine-2-carbonitrile
CC1=C(C=CC(=N1)C#N)N
Quantity 250mg
UN Number UN3439
Beilstein 15464648
Formula Weight 133.15
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-Amino-6-cyano-2-methylpyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.