Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-6-morpholinopyridazine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
66346-91-6
C8H12N4O
180.21
IEUHTZVAUDMKQJ-UHFFFAOYSA-N
6-morpholinopyridazin-3-amine,3-amino-6-morpholin-4-yl pyridazine,6-morpholin-4-yl pyridazin-3-amine,3-amino-6-morpholin-4-ylpyridazine,6-morpholin-4-yl-pyridazin-3-ylamine,3-pyridazinamine, 6-4-morpholinyl,6-morpholinopyridazine-3-amine,6-4-morpholinyl-3-pyridazinamine,3-amino-6-morpholin-4-3-amino-6-morpholin-4-ylpyridazine
13114295
6-morpholin-4-ylpyridazin-3-amine
C1COCCN1C2=NN=C(C=C2)N
66346-91-6
C8H12N4O
180.21
IEUHTZVAUDMKQJ-UHFFFAOYSA-N
6-morpholinopyridazin-3-amine,3-amino-6-morpholin-4-yl pyridazine,6-morpholin-4-yl pyridazin-3-amine,3-amino-6-morpholin-4-ylpyridazine,6-morpholin-4-yl-pyridazin-3-ylamine,3-pyridazinamine, 6-4-morpholinyl,6-morpholinopyridazine-3-amine,6-4-morpholinyl-3-pyridazinamine,3-amino-6-morpholin-4-3-amino-6-morpholin-4-ylpyridazine
13114295
6-morpholin-4-ylpyridazin-3-amine
C1COCCN1C2=NN=C(C=C2)N
Melting Point 181°C to 183°C
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 180.21
Physical Form Crystalline Powder or Powder
Percent Purity 97%
Chemical Name or Material 3-Amino-6-morpholinopyridazine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning