Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-9-ethylcarbazole, 90%, tech., ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
132-32-1
C14H14N2
210.28
MFCD00004964
OXEUETBFKVCRNP-UHFFFAOYSA-N
3-amino-9-ethylcarbazole,9-ethyl-9h-carbazol-3-amine,3-amino-n-ethylcarbazole,9h-carbazol-3-amine, 9-ethyl,carbazole, 3-amino-9-ethyl,9-ethylcarbazol-3-ylamine,unii-8q2bg27jbu,9-ethylcarbazole-3-ylamine,8q2bg27jbu,9-ethyl-9h-carbazol-3-ylamine
8588
9-ethylcarbazol-3-amine
CCN1C2=C(C=C(C=C2)N)C3=CC=CC=C31
132-32-1
C14H14N2
210.28
MFCD00004964
OXEUETBFKVCRNP-UHFFFAOYSA-N
3-amino-9-ethylcarbazole,9-ethyl-9h-carbazol-3-amine,3-amino-n-ethylcarbazole,9h-carbazol-3-amine, 9-ethyl,carbazole, 3-amino-9-ethyl,9-ethylcarbazol-3-ylamine,unii-8q2bg27jbu,9-ethylcarbazole-3-ylamine,8q2bg27jbu,9-ethyl-9h-carbazol-3-ylamine
8588
9-ethylcarbazol-3-amine
CCN1C2=C(C=C(C=C2)N)C3=CC=CC=C31
Packaging Glass bottle
Melting Point 98.0°C to 100.0°C
Quantity 5g
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Beilstein 22, IV, 4936
Solubility Information Solubility in water: insoluble.
Formula Weight 210.28
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Amino-9-ethylcarbazole
Show More Show Less

GHS H Statement
Toxic if swallowed.
May cause cancer.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
Wear protective gloves/protective clothing/e

GHS Signal Word: Danger

EINECSNumber : 205-057-7

RTECSNumber : FE3590000

TSCA : TSCA