Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-N-methylbenzylamine 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
18759-96-1
C8H12N2
136.198
MFCD00211072
HBLPYXIZPMDWIO-UHFFFAOYSA-N
3-(Methylaminomethyl)aniline
11643936
3-(methylaminomethyl)aniline
CNCC1=CC(=CC=C1)N
18759-96-1
C8H12N2
136.198
MFCD00211072
HBLPYXIZPMDWIO-UHFFFAOYSA-N
3-(Methylaminomethyl)aniline
11643936
3-(methylaminomethyl)aniline
CNCC1=CC(=CC=C1)N
Color White-Yellow
Quantity 25g
UN Number 3259
Formula Weight 136.20
Physical Form Crystal-Lump at 20°C
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 3-Amino-N-methylbenzylamine