Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-o-cresol, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
53222-92-7
C7H9NO
123.15
MFCD00007787
FLROJJGKUKLCAE-UHFFFAOYSA-N
3-amino-o-cresol,phenol, 3-amino-2-methyl,2-amino-o-cresol,2-methyl-3-aminophenol,3-hydroxy-2-methylaniline,aminokresol,o-cresol, 3-amino,pubchem15039,2-methyl-3-amino-phenol,3-amino-2-methyl phenol
104448
3-amino-2-methylphenol
CC1=C(C=CC=C1O)N
53222-92-7
C7H9NO
123.15
MFCD00007787
FLROJJGKUKLCAE-UHFFFAOYSA-N
3-amino-o-cresol,phenol, 3-amino-2-methyl,2-amino-o-cresol,2-methyl-3-aminophenol,3-hydroxy-2-methylaniline,aminokresol,o-cresol, 3-amino,pubchem15039,2-methyl-3-amino-phenol,3-amino-2-methyl phenol
104448
3-amino-2-methylphenol
CC1=C(C=CC=C1O)N
Melting Point 127°C to 133°C
Quantity 250mg
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula H2NC6H3(CH3)OH
Beilstein 13, 579
Formula Weight 123.15
Percent Purity 95%
Chemical Name or Material 3-Amino-o-cresol, 95%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 258-438-5

TSCA : TSCA