Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3′-Aminoacetophenone 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
99-03-6
C8H9NO
135.166
MFCD00007796
CKQHAYFOPRIUOM-UHFFFAOYSA-N
3'-aminoacetophenone,1-3-aminophenyl ethanone,m-aminoacetophenone,3-aminoacetophenone,3-acetylaniline,m-acetylaniline,ethanone, 1-3-aminophenyl,1-3-aminophenyl ethan-1-one,m-aminoacetylbenzene,acetophenone, 3'-amino
7417
1-(3-aminophenyl)ethanone
CC(=O)C1=CC(=CC=C1)N
99-03-6
C8H9NO
135.166
MFCD00007796
CKQHAYFOPRIUOM-UHFFFAOYSA-N
3'-aminoacetophenone,1-3-aminophenyl ethanone,m-aminoacetophenone,3-aminoacetophenone,3-acetylaniline,m-acetylaniline,ethanone, 1-3-aminophenyl,1-3-aminophenyl ethan-1-one,m-aminoacetylbenzene,acetophenone, 3'-amino
7417
1-(3-aminophenyl)ethanone
CC(=O)C1=CC(=CC=C1)N
Quantity 500g
Boiling Point 252°C
Color Undesignated
Melting Point 98°C
Formula Weight 135.17
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3′-Aminoacetophenone

EINECSNumber : (3)-1242

RTECSNumber : AM5800000

TSCA : Yes