Learn More
3-Aminobenzamide, 98%
Description
3-Aminobenzamide is used as an endogenous poly-ADP-ribosyltransferase inhibitor. Through its effects on PARPs, 3-amino benzamide causes telomere shortening and stimulates angiogenesis. It displays neuroprotective, anti-necrotic effects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Aminobenzamide is used as an endogenous poly-ADP-ribosyltransferase inhibitor. Through its effects on PARPs, 3-amino benzamide causes telomere shortening and stimulates angiogenesis. It displays neuroprotective, anti-necrotic effects.
Solubility
Soluble in dimethylformamide (∼30 mg/ml), dimethyl sulfoxide (∼30 mg/ml), phosphate buffered saline (pH7.2) (∼2 mg/ml), water (25 mg/ml), and ethanol (25 mg/ml).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 111°C to 115°C |
Quantity | 5 g |
Beilstein | 2802373 |
Solubility Information | Soluble in dimethylformamide (∽30mg/ml),dimethyl sulfoxide (∽30mg/ml),phosphate buffered saline (pH7.2) (∽2mg/ml),water (25mg/ml),and ethanol (25mg/ml). |
Formula Weight | 136.15 |
Percent Purity | 98% |
Chemical Name or Material | 3-Aminobenzamide |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 222-586-9 |
RTECSNumber | CU8992000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.