Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminobenzeneboronic acid hemisulfate, 98+%

Click to view available options
Quantity:
1g
5g
25g
66472-86-4
C12H18B2N2O8S
371.962
MFCD00013111
UKTAURVTSWDIQR-UHFFFAOYSA-N
3-aminophenyl boronic acid sulfate 2:1,3-aminobenzeneboronic acid hemisulfate salt,3-aminophenylboronic acid hemisulfate,3-aminobenzeneboronic acid hemisulfate,3-aminophenylboronic acid hemisulphate,4-aminophenylboronic acid hemisulfate,m-aminophenyl boronic acid, hemisulphate,boronic acid, 3-aminophenyl-, sulfate 2:1,3-aminophenyl boronic acid; sulfuric acid,bis m-aminophenylboronic acid ; sulfuric acid
16211139
(3-aminophenyl)boronic acid;sulfuric acid
B(C1=CC(=CC=C1)N)(O)O.B(C1=CC(=CC=C1)N)(O)O.OS(=O)(=O)O
66472-86-4
C12H18B2N2O8S
371.962
MFCD00013111
UKTAURVTSWDIQR-UHFFFAOYSA-N
3-aminophenyl boronic acid sulfate 2:1,3-aminobenzeneboronic acid hemisulfate salt,3-aminophenylboronic acid hemisulfate,3-aminobenzeneboronic acid hemisulfate,3-aminophenylboronic acid hemisulphate,4-aminophenylboronic acid hemisulfate,m-aminophenyl boronic acid, hemisulphate,boronic acid, 3-aminophenyl-, sulfate 2:1,3-aminophenyl boronic acid; sulfuric acid,bis m-aminophenylboronic acid ; sulfuric acid
16211139
(3-aminophenyl)boronic acid;sulfuric acid
B(C1=CC(=CC=C1)N)(O)O.B(C1=CC(=CC=C1)N)(O)O.OS(=O)(=O)O
Quantity 25g
Sensitivity Hygroscopic
Melting Point >300°C
Beilstein 8094151
Formula Weight 185.98
Percent Purity ≥98%
Chemical Name or Material 3-Aminobenzeneboronic acid hemisulfate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 266-376-5

TSCA : No

Recommended Storage : Ambient temperatures