Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminobenzenesulfonamide, 97+%

Click to view available options
Quantity:
5g
25g
98-18-0
C6H8N2O2S
172.202
MFCD00035781
JPVKCHIPRSQDKL-UHFFFAOYSA-N
metanilamide,m-aminobenzenesulfonamide,benzenesulfonamide, 3-amino,m-sulfamoylaniline,3-aminobenzene-1-sulfonamide,benzenesulfonamide, m-amino,m-aminobenzenesulphonamide,metaniilamide,3-aminosulfonylaniline,3-amino-benzenesulfonamide
7377
3-aminobenzenesulfonamide
C1=CC(=CC(=C1)S(=O)(=O)N)N
98-18-0
C6H8N2O2S
172.202
MFCD00035781
JPVKCHIPRSQDKL-UHFFFAOYSA-N
metanilamide,m-aminobenzenesulfonamide,benzenesulfonamide, 3-amino,m-sulfamoylaniline,3-aminobenzene-1-sulfonamide,benzenesulfonamide, m-amino,m-aminobenzenesulphonamide,metaniilamide,3-aminosulfonylaniline,3-amino-benzenesulfonamide
7377
3-aminobenzenesulfonamide
C1=CC(=CC(=C1)S(=O)(=O)N)N
Quantity 5g
Melting Point 139°C to 142°C
Formula Weight 172.21
Percent Purity ≥97%
Chemical Name or Material 3-Aminobenzenesulfonamide

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 202-646-0

RTECSNumber : OY2200000

TSCA : Yes

Recommended Storage : Ambient temperatures