Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzenesulfonamide 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
98-18-0
C6H8N2O2S
172.202
MFCD00035781
JPVKCHIPRSQDKL-UHFFFAOYSA-N
metanilamide,m-aminobenzenesulfonamide,benzenesulfonamide, 3-amino,m-sulfamoylaniline,3-aminobenzene-1-sulfonamide,benzenesulfonamide, m-amino,m-aminobenzenesulphonamide,metaniilamide,3-aminosulfonylaniline,3-amino-benzenesulfonamide
7377
3-aminobenzenesulfonamide
C1=CC(=CC(=C1)S(=O)(=O)N)N
98-18-0
C6H8N2O2S
172.202
MFCD00035781
JPVKCHIPRSQDKL-UHFFFAOYSA-N
metanilamide,m-aminobenzenesulfonamide,benzenesulfonamide, 3-amino,m-sulfamoylaniline,3-aminobenzene-1-sulfonamide,benzenesulfonamide, m-amino,m-aminobenzenesulphonamide,metaniilamide,3-aminosulfonylaniline,3-amino-benzenesulfonamide
7377
3-aminobenzenesulfonamide
C1=CC(=CC(=C1)S(=O)(=O)N)N
Color Undesignated
Quantity 250g
Melting Point 142°C
Formula Weight 172.20
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 3-Aminobenzenesulfonamide

EINECSNumber : (3)-2179

RTECSNumber : OY2200000

TSCA : Yes