Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminobenzenesulfonamide, tech. 80%

Manufacturer:  Alfa Aesar™ B2543922

Quantity 100g

 View more versions of this product

Catalog No. AAB2543922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

98-18-0
172.21
Metanilamide; Metanilamide
Technical Grade
C6H8N2O2S
MFCD00035781
3-Aminobenzenesulfonamide

Specifications

98-18-0
C6H8N2O2S
100g
Metanilamide; Metanilamide
80%
139° to 142°C
172.21
MFCD00035781
3-Aminobenzenesulfonamide
Technical Grade

Safety and Handling

SDS & Safety Documents

RTECSNumber : OY2200000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit