Alfa Aesar™ 3-Aminobenzenesulfonamide, tech. 80%

Manufacturer: Alfa Aesar™ B2543936

Quantity 500g

 View more versions of this product

Catalog No. AAB2543936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98-18-0
    Molecular Formula C6H8N2O2S
    Formula Weight 172.21
    MDL Number MFCD00035781
    Synonym Metanilamide; Metanilamide
    Chemical Name or Material 3-Aminobenzenesulfonamide
    Grade Technical Grade

    Specifications

    Formula Weight 172.21
    Melting Point 139° to 142°C
    Molecular Formula C6H8N2O2S
    Quantity 500g
    CAS 98-18-0
    RTECS Number OY2200000
    TSCA TSCA
    MDL Number MFCD00035781
    Synonym Metanilamide; Metanilamide
    Chemical Name or Material 3-Aminobenzenesulfonamide
    Assay 80%
    Grade Technical Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit