Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzonitrile, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2237-30-1
C7H6N2
118.14
MFCD00007756
NJXPYZHXZZCTNI-UHFFFAOYSA-N
3-cyanoaniline,m-aminobenzonitrile,m-cyanoaniline,3-amino-benzonitrile,3-aminobenzonitril,benzonitrile, m-amino,m-anthranilonitrile,benzonitrile, 3-amino,unii-15v3cw939x,3-amino benzonitrile
16702
3-aminobenzonitrile
C1=CC(=CC(=C1)N)C#N
2237-30-1
C7H6N2
118.14
MFCD00007756
NJXPYZHXZZCTNI-UHFFFAOYSA-N
3-cyanoaniline,m-aminobenzonitrile,m-cyanoaniline,3-amino-benzonitrile,3-aminobenzonitril,benzonitrile, m-amino,m-anthranilonitrile,benzonitrile, 3-amino,unii-15v3cw939x,3-amino benzonitrile
16702
3-aminobenzonitrile
C1=CC(=CC(=C1)N)C#N
Packaging Glass bottle
Quantity 5g
Color Brown
Flash Point 112°C
Melting Point 50.0°C to 54.0°C
Infrared Spectrum Authentic
Boiling Point 288.0°C to 290.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula H2NC6H4CN
Beilstein 14, 391
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ehtanol,ethyl ether,acetone
Formula Weight 118.14
Physical Form Crystalline Solid
Percent Purity 99%
Chemical Name or Material 3-Aminobenzonitrile, 99%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POI

GHS Signal Word: Danger

EINECSNumber : 218-800-5

RTECSNumber : DI2454000

TSCA : TSCA