Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzophenone 98.0+%, TCI America™

Click to view available options
Quantity:
5g
2835-78-1
C13H11NO
197.237
FUADXEJBHCKVBN-UHFFFAOYSA-N
3-aminobenzophenone,3-benzoylaniline,methanone, 3-aminophenyl phenyl,3-aminophenyl phenyl methanone,benzophenone, 3-amino,m-aminobenzophenone,3-aminophenyl phenylmethanone,3-aminophenyl phenyl ketone,3-amino-benzophenon,3-amino-benzophenone
17817
(3-aminophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)N
2835-78-1
C13H11NO
197.237
FUADXEJBHCKVBN-UHFFFAOYSA-N
3-aminobenzophenone,3-benzoylaniline,methanone, 3-aminophenyl phenyl,3-aminophenyl phenyl methanone,benzophenone, 3-amino,m-aminobenzophenone,3-aminophenyl phenylmethanone,3-aminophenyl phenyl ketone,3-amino-benzophenon,3-amino-benzophenone
17817
(3-aminophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)N
Melting Point 86°C
Quantity 5g
Color Yellow
Formula Weight 197.24
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 3-Aminobenzophenone

RTECSNumber : DJ0170000

TSCA : No