Alfa Aesar™ 3-Aminobiphenyl hydrochloride

Manufacturer: Alfa Aesar™ H5102706

Quantity 5g

 View more versions of this product

Catalog No. AAH5102706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2113-55-5
    Molecular Formula C12H11N·HCl
    Formula Weight 205.69
    MDL Number MFCD00068358
    Synonym 3-Biphenylamine hydrochloride; 3-Phenylaniline hydrochloride
    Chemical Name or Material 3-Aminobiphenyl hydrochloride

    Specifications

    Formula Weight 205.69
    Molecular Formula C12H11N·HCl
    Quantity 5g
    MDL Number MFCD00068358
    Odor Odorless
    Synonym 3-Biphenylamine hydrochloride; 3-Phenylaniline hydrochloride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H351-H302
    Suspected of causing cancer.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280h
    Wear protective gloves/clothing.
    CAS 2113-55-5
    Chemical Name or Material 3-Aminobiphenyl hydrochloride

    GHS H Statement
    H351-H302
    Suspected of causing cancer.
    Harmful if swallowed.

    GHS P Statement
    P280h
    Wear protective gloves/clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit