Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminocrotononitrile, 96%, mixture of cis and trans, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
1118-61-2
C4H6N2
82.1
MFCD00008071
DELJOESCKJGFML-RQOWECAXSA-N
3-aminocrotononitrile,2z-3-aminobut-2-enenitrile,3-amino-2-butenenitrile,z-3-aminobut-2-enenitrile,beta-aminocrotononitrile,3-amino-2-butenonitrile,2-amino-1-propenecarbonitrile,3-amino-crotononitrile,beta-amino-crotononitrile,2z-3-amino-2-butenenitrile
5325263
(Z)-3-aminobut-2-enenitrile
CC(=CC#N)N
1118-61-2
C4H6N2
82.1
MFCD00008071
DELJOESCKJGFML-RQOWECAXSA-N
3-aminocrotononitrile,2z-3-aminobut-2-enenitrile,3-amino-2-butenenitrile,z-3-aminobut-2-enenitrile,beta-aminocrotononitrile,3-amino-2-butenonitrile,2-amino-1-propenecarbonitrile,3-amino-crotononitrile,beta-amino-crotononitrile,2z-3-amino-2-butenenitrile
5325263
(Z)-3-aminobut-2-enenitrile
CC(=CC#N)N
Packaging Plastic bottle
Infrared Spectrum Authentic
Quantity 100g
Color Yellow
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula CH3C(NH2)=CHCN
Beilstein 03, 660
Formula Weight 82.1
Physical Form Flakes
Percent Purity 96%
Chemical Name or Material 3-Aminocrotononitrile, 96%
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Cal

GHS Signal Word: Warning

EINECSNumber : 214-266-2

TSCA : TSCA