Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminoisonicotinic acid, 95+%, Maybridge

Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
7579-20-6
C6H6N2O2
138.126
FYEQKMAVRYRMBL-UHFFFAOYSA-N
3-aminoisonicotinic acid,3-amino-4-pyridinecarboxylic acid,3-amino-isonicotinic acid,3-amino-4-pyridine carboxylic acid,4-pyridinecarboxylic acid, 3-amino,3-amino-4-carboxypyridine,3-amino-4-pyridinecarboxylicacid,3-amino-4-carboxylic acid pyridine,pubchem2420,3-aminoisonicotinsaure
459531
3-aminopyridine-4-carboxylic acid
C1=CN=CC(=C1C(=O)O)N
7579-20-6
C6H6N2O2
138.126
FYEQKMAVRYRMBL-UHFFFAOYSA-N
3-aminoisonicotinic acid,3-amino-4-pyridinecarboxylic acid,3-amino-isonicotinic acid,3-amino-4-pyridine carboxylic acid,4-pyridinecarboxylic acid, 3-amino,3-amino-4-carboxypyridine,3-amino-4-pyridinecarboxylicacid,3-amino-4-carboxylic acid pyridine,pubchem2420,3-aminoisonicotinsaure
459531
3-aminopyridine-4-carboxylic acid
C1=CN=CC(=C1C(=O)O)N
Packaging Amber Glass Bottle
Quantity 1g
Assay Percent Range 95%
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material 3-Aminoisonicotinic acid