Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminoisoxazole, 95%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
1750-42-1
C3H4N2O
84.08
MFCD00038814
RHFWLPWDOYJEAL-UHFFFAOYSA-N
3-aminoisoxazole,isoxazol-3-amine,3-isoxazolamine,3-amino isoxazole,isoxazolamine,3-amineisoxazole,aminoisoxazole,3-aminoisoxazol,3-amino-1,2-oxazole,amino-isoxazole
1810216
1,2-oxazol-3-amine
C1=CON=C1N
1750-42-1
C3H4N2O
84.08
MFCD00038814
RHFWLPWDOYJEAL-UHFFFAOYSA-N
3-aminoisoxazole,isoxazol-3-amine,3-isoxazolamine,3-amino isoxazole,isoxazolamine,3-amineisoxazole,aminoisoxazole,3-aminoisoxazol,3-amino-1,2-oxazole,amino-isoxazole
1810216
1,2-oxazol-3-amine
C1=CON=C1N
Flash Point >110°C
Boiling Point 226°C to 228°C
Specific Gravity 1.38
Quantity 5mL
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Density 1.3800g/mL
Refractive Index 1.5100 to 1.5120 (20°C, 589nm)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 84.08
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 3-Aminoisoxazole
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning