Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Aminomethyl)-1-methylindole, 95%, Acros Organics™

Click to view available options
Quantity:
5g
19293-60-8
C10H12N2
160.22
NOFZMDGMQKRLIV-UHFFFAOYSA-N
1-methyl-1h-indol-3-yl methanamine,1-methylindol-3-yl methanamine,1-methyl-3-indolemethylamine,1-methyl-1h-indol-3-yl-methylamine,1-methyl-1h-indol-3-yl methylamine,1-1-methyl-1h-indol-3-yl methanamine,c-1-methyl-1h-indol-3-yl-methylamine,1-1-methylindol-3-yl methanamine,n-methyl-3-aminomethyl-indol,1-methyl-1h-indole-3-methanamine
3934542
(1-methylindol-3-yl)methanamine
CN1C=C(C2=CC=CC=C21)CN
19293-60-8
C10H12N2
160.22
NOFZMDGMQKRLIV-UHFFFAOYSA-N
1-methyl-1h-indol-3-yl methanamine,1-methylindol-3-yl methanamine,1-methyl-3-indolemethylamine,1-methyl-1h-indol-3-yl-methylamine,1-methyl-1h-indol-3-yl methylamine,1-1-methyl-1h-indol-3-yl methanamine,c-1-methyl-1h-indol-3-yl-methylamine,1-1-methylindol-3-yl methanamine,n-methyl-3-aminomethyl-indol,1-methyl-1h-indole-3-methanamine
3934542
(1-methylindol-3-yl)methanamine
CN1C=C(C2=CC=CC=C21)CN
Quantity 5g
Infrared Spectrum Authentic
Formula Weight 160.22
Percent Purity ≥94.0% (HPLC)
Chemical Name or Material 3-(Aminomethyl)-1-methylindole

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

Warning