Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminomethyl-2-bromothiophene, 95%

Click to view available options
Quantity:
250mg
1g
157664-47-6
C5H6BrNS
192.074
MFCD02180914
SLKZUJBSIKGHFU-UHFFFAOYSA-N
2-bromo-3-thienyl methylamine,2-bromothiophen-3-yl methanamine,2-bromo-3-thiophenemethylamine,3-thiophenemethanamine,2-bromo,1-2-bromothiophen-3-yl methanamine,2-bromothien-3-yl methylamine hydrochloride,pubchem5276,h-val-ssna,bestipharma 583-851,3-aminomethyl-2-bromothiophene
2735586
(2-bromothiophen-3-yl)methanamine
C1=CSC(=C1CN)Br
157664-47-6
C5H6BrNS
192.074
MFCD02180914
SLKZUJBSIKGHFU-UHFFFAOYSA-N
2-bromo-3-thienyl methylamine,2-bromothiophen-3-yl methanamine,2-bromo-3-thiophenemethylamine,3-thiophenemethanamine,2-bromo,1-2-bromothiophen-3-yl methanamine,2-bromothien-3-yl methylamine hydrochloride,pubchem5276,h-val-ssna,bestipharma 583-851,3-aminomethyl-2-bromothiophene
2735586
(2-bromothiophen-3-yl)methanamine
C1=CSC(=C1CN)Br
Density 1.657g/mL
Sensitivity Air Sensitive
Refractive Index 1.6100
Quantity 1g
Color Colorless to Yellow
UN Number UN2810
Formula Weight 192.08
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 3-Aminomethyl-2-bromothiophene

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.