Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Aminomethyl)benzoic acid hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
876-03-9
C8H10ClNO2
187.623
MFCD03791117
SJCCOASSOPUHEN-UHFFFAOYSA-N
3-aminomethyl benzoic acid hydrochloride,3-aminomethylbenzoic acid hydrochloride,3-aminomethylbenzoic acid hcl,3-aminomethyl-benzoic acid hydrochloride,acmc-209tzp,3-aminomethyl benzoic acid hcl,3-carboxybenzylamine hydrochloride,3-aminomethyl-benzoic acid hcl salt,3-aminomethyl benzoic acid, chloride,3-aminomethyl benzoicacid hydrochloride
16218772
3-(aminomethyl)benzoic acid;hydrochloride
C1=CC(=CC(=C1)CN)C(=O)O.Cl
876-03-9
C8H10ClNO2
187.623
MFCD03791117
SJCCOASSOPUHEN-UHFFFAOYSA-N
3-aminomethyl benzoic acid hydrochloride,3-aminomethylbenzoic acid hydrochloride,3-aminomethylbenzoic acid hcl,3-aminomethyl-benzoic acid hydrochloride,acmc-209tzp,3-aminomethyl benzoic acid hcl,3-carboxybenzylamine hydrochloride,3-aminomethyl-benzoic acid hcl salt,3-aminomethyl benzoic acid, chloride,3-aminomethyl benzoicacid hydrochloride
16218772
3-(aminomethyl)benzoic acid;hydrochloride
C1=CC(=CC(=C1)CN)C(=O)O.Cl
Applications
It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Melting Point 242°C to 244°C
Quantity 5g
Beilstein 3697656
Formula Weight 187.62
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-(Aminomethyl)benzoic acid hydrochloride

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C