Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminonoradamantane hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
86128-83-8
C9H16ClN
173.684
MFCD00213499
YPEGOYFGBDTDJK-UHFFFAOYSA-N
3-noradamantanamine hydrochloride,octahydro-2,5-methanopentalen-3a-amine hydrochloride,3-aminonoradamantane hcl,3-aminonoradamantane hydrochloride,tricyclo 3.3.1.0 3 ,? nonan-3-amine hydrochloride,3-noradamantanamine hcl,c9h15n.hcl,3-noradamantanaminehydrochloride,3-noradamantanamine hydrochloride 95,3-noradamantanamine hydrochloride
12791864
C1C2CC3CC1CC3(C2)N.Cl
86128-83-8
C9H16ClN
173.684
MFCD00213499
YPEGOYFGBDTDJK-UHFFFAOYSA-N
3-noradamantanamine hydrochloride,octahydro-2,5-methanopentalen-3a-amine hydrochloride,3-aminonoradamantane hcl,3-aminonoradamantane hydrochloride,tricyclo 3.3.1.0 3 ,? nonan-3-amine hydrochloride,3-noradamantanamine hcl,c9h15n.hcl,3-noradamantanaminehydrochloride,3-noradamantanamine hydrochloride 95,3-noradamantanamine hydrochloride
12791864
C1C2CC3CC1CC3(C2)N.Cl
Color White
Melting Point 350°C
Quantity 250mg
Formula Weight 173.68
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 3-Aminonoradamantane hydrochloride