Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminooxetane, 95%, Acros Organics

Click to view available options
Quantity:
250mg
500mg
21635-88-1
C3H7NO
73.09
OJEOJUQOECNDND-UHFFFAOYSA-N
3-oxetanamine,3-aminooxetane,oxetan-3-ylamine,3-oxetanamin,3-amino-oxetane,oxetane-3-amine,oxetan-3-yl amine,pubchem20414,acmc-1cgam
9833923
oxetan-3-amine
C1C(CO1)N
21635-88-1
C3H7NO
73.09
OJEOJUQOECNDND-UHFFFAOYSA-N
3-oxetanamine,3-aminooxetane,oxetan-3-ylamine,3-oxetanamin,3-amino-oxetane,oxetane-3-amine,oxetan-3-yl amine,pubchem20414,acmc-1cgam
9833923
oxetan-3-amine
C1C(CO1)N
Boiling Point 99°C
Flash Point 24°C
Quantity 250mg
Assay Percent Range 94% min. (GC)
Formula Weight 73.09
Percent Purity 95%
Chemical Name or Material 3-Aminooxetane

GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water f

GHS Signal Word: Danger