Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopentane, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
616-24-0
C5H13N
87.16
MFCD00008096
PQPFFKCJENSZKL-UHFFFAOYSA-N
3-aminopentane,3-pentanamine,1-ethylpropylamine,3-pentylamine,propylamine, 1-ethyl,3-amylamine,unii-3n2it605hv,1-ethyl-propylamine,pent-3-ylamine,3-amino pentane
12019
CHEBI:84248
pentan-3-amine
CCC(CC)N
616-24-0
C5H13N
87.16
MFCD00008096
PQPFFKCJENSZKL-UHFFFAOYSA-N
3-aminopentane,3-pentanamine,1-ethylpropylamine,3-pentylamine,propylamine, 1-ethyl,3-amylamine,unii-3n2it605hv,1-ethyl-propylamine,pent-3-ylamine,3-amino pentane
12019
CHEBI:84248
pentan-3-amine
CCC(CC)N
Specific Gravity 0.74
Packaging Glass bottle
Density 0.7400g/mL
Quantity 100g
Boiling Point 89.0°C to 91.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Flash Point 2°C
Refractive Index 1.4055
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula (C2H5)2CHNH2
Beilstein 04, 179
Solubility Information Solubility in water: soluble.
Formula Weight 87.16
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 3-Aminopentane
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 210-471-6