Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminophenol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
Packaging:
Glass bottle
591-27-5
C6H7NO
109.13
MFCD00007786
CWLKGDAVCFYWJK-UHFFFAOYSA-N
m-aminophenol,m-hydroxyaniline,3-hydroxyaniline,phenol, 3-amino,1-amino-3-hydroxybenzene,3-amino-1-hydroxybenzene,fouramine eg,futramine eg,fourrine eg,pelagol eg
11568
CHEBI:28924
3-aminophenol
C1=CC(=CC(=C1)O)N
591-27-5
C6H7NO
109.13
MFCD00007786
CWLKGDAVCFYWJK-UHFFFAOYSA-N
m-aminophenol,m-hydroxyaniline,3-hydroxyaniline,phenol, 3-amino,1-amino-3-hydroxybenzene,3-amino-1-hydroxybenzene,fouramine eg,futramine eg,fourrine eg,pelagol eg
11568
CHEBI:28924
3-aminophenol
C1=CC(=CC(=C1)O)N
Boiling Point 164°C (15.0 hPa)
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 121.0°C to 124.0°C
Flash Point 155°C
Color Gray or Pink to White
Quantity 250g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula H2NC6H4OH
Beilstein 13, 401
Merck Index 15, 456
Solubility Information Solubility in water: 35g/L (20°C). Other solubilities: freely soluble in hot water, freely soluble in alcohol, ether and amyl alcohol, slightly soluble in benzene, very slightly soluble in petroleum ether
Formula Weight 109.13
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 3-Aminophenol, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 209-711-2

RTECSNumber : SJ4900000

TSCA : TSCA