Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminophenylsulfur Pentafluoride 95.0+%, TCI America™

Click to view available options
5G
2993-22-8
C6H6F5NS
219.173
MFCD00674199
NUFLICUHOXHWER-UHFFFAOYSA-N
3-aminophenylsulfur pentafluoride,3-pentafluorothio aniline,3-pentafluorosulfanyl aniline,3-aminophenylsulphur pentafluoride,sulfur, 3-aminophenyl pentafluoro-, oc-6-21-9ci,3-pentafluoro-??-sulfanyl aniline,acmc-1cou4,3-pentafluorosulfanylaniline,3-pentafluorosulphanylaniline,3-pentafluorosulfur aniline
2779188
3-(pentafluoro-$l^{6}-sulfanyl)aniline
C1=CC(=CC(=C1)S(F)(F)(F)(F)F)N
2993-22-8
C6H6F5NS
219.173
MFCD00674199
NUFLICUHOXHWER-UHFFFAOYSA-N
3-aminophenylsulfur pentafluoride,3-pentafluorothio aniline,3-pentafluorosulfanyl aniline,3-aminophenylsulphur pentafluoride,sulfur, 3-aminophenyl pentafluoro-, oc-6-21-9ci,3-pentafluoro-??-sulfanyl aniline,acmc-1cou4,3-pentafluorosulfanylaniline,3-pentafluorosulphanylaniline,3-pentafluorosulfur aniline
2779188
3-(pentafluoro-$l^{6}-sulfanyl)aniline
C1=CC(=CC(=C1)S(F)(F)(F)(F)F)N
Color Yellow
Melting Point 35°C
Quantity 5g
Boiling Point 87°C
UN Number 2811
Formula Weight 219.17
Physical Form Crystal-Lump at 20°C
Percent Purity ≥95.0% (T)
Chemical Name or Material 3-Aminophenylsulfur Pentafluoride

TSCA : No

Recommended Storage : Refrigerator