Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopropyltriethoxysilane, 99%, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
919-30-2
C9H23NO3Si
221.37
MFCD00008207
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-aminopropyltriethoxysilane,3-aminopropyl triethoxysilane,aptes,3-triethoxysilyl propan-1-amine,1-propanamine, 3-triethoxysilyl,silicone a-1100,silane 1100,3-triethoxysilyl propylamine,propylamine, 3-triethoxysilyl,triethoxy 3-aminopropyl silane
13521
3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC
919-30-2
C9H23NO3Si
221.37
MFCD00008207
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-aminopropyltriethoxysilane,3-aminopropyl triethoxysilane,aptes,3-triethoxysilyl propan-1-amine,1-propanamine, 3-triethoxysilyl,silicone a-1100,silane 1100,3-triethoxysilyl propylamine,propylamine, 3-triethoxysilyl,triethoxy 3-aminopropyl silane
13521
3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC
Specific Gravity 0.942
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Melting Point -70.0°C
Flash Point 96°C
Boiling Point 217.0°C
Quantity 100mL
Color Undesignated
Refractive Index 1.4200 to 1.4220
Density 0.9420g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula H2N(CH2)3Si(OC2H5)3
Solubility Information Solubility in water: reacts. Other solubilities: soluble in chloroform
Formula Weight 221.37
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Aminopropyltriethoxysilane
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 213-048-4