Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminopyrazine-2-carboxylic acid, 98+%

Click to view available options
Quantity:
1g
5g
25g
5424-01-1
C5H5N3O2
139.11
MFCD00006141
ZAGZIOYVEIDDJA-UHFFFAOYSA-N
3-amino-2-pyrazinecarboxylic acid,pyrazinecarboxylic acid, 3-amino,2-amino-3-pyrazinecarboxylic acid,3-aminopiperazine-2-carboxylic acid,2-aminopyrazine-3-carboxylic acid,2-pyrazinecarboxylic acid, 3-amino,3-aminopyrazine-2-carboxyliacid,zlchem 459,pubchem8544,3-aminopyrazinoic acid
72656
3-aminopyrazine-2-carboxylic acid
C1=CN=C(C(=N1)C(=O)O)N
5424-01-1
C5H5N3O2
139.11
MFCD00006141
ZAGZIOYVEIDDJA-UHFFFAOYSA-N
3-amino-2-pyrazinecarboxylic acid,pyrazinecarboxylic acid, 3-amino,2-amino-3-pyrazinecarboxylic acid,3-aminopiperazine-2-carboxylic acid,2-aminopyrazine-3-carboxylic acid,2-pyrazinecarboxylic acid, 3-amino,3-aminopyrazine-2-carboxyliacid,zlchem 459,pubchem8544,3-aminopyrazinoic acid
72656
3-aminopyrazine-2-carboxylic acid
C1=CN=C(C(=N1)C(=O)O)N
Quantity 25g
Melting Point 204°C to 206°C (decomposition)
Beilstein 124835
Formula Weight 139.11
Percent Purity ≥98%
Chemical Name or Material 3-Aminopyrazine-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning