Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminopyrazole-4-carboxamidoxime, 97%

Click to view available options
Quantity:
250mg
1g
58046-48-3
58046-48-3
Quantity 1g
Color White
Formula Weight 141.13
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Aminopyrazole-4-carboxamidoxime