Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminopyrazole-4-thiocarboxamide, 97%

Click to view available options
Quantity:
250mg
1g
34954-13-7
C4H6N4S
142.18
VRWNWVXHINBCSC-UHFFFAOYSA-N
3-amino-1h-pyrazole-4-carbothioamide,3-amino-1h-pyrazole-4-thiocarboxamide,amino 5-aminopyrazol-4-yl methane-1-thione,1h-pyrazole-4-carbothioamide, 5-amino,3-amino-2h-pyrazole-4-carbothioamide
8027117
5-amino-1H-pyrazole-4-carbothioamide
C1=NNC(=C1C(=S)N)N
34954-13-7
C4H6N4S
142.18
VRWNWVXHINBCSC-UHFFFAOYSA-N
3-amino-1h-pyrazole-4-carbothioamide,3-amino-1h-pyrazole-4-thiocarboxamide,amino 5-aminopyrazol-4-yl methane-1-thione,1h-pyrazole-4-carbothioamide, 5-amino,3-amino-2h-pyrazole-4-carbothioamide
8027117
5-amino-1H-pyrazole-4-carbothioamide
C1=NNC(=C1C(=S)N)N
Quantity 250mg
Color Brown to White
Formula Weight 142.18
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Aminopyrazole-4-thiocarboxamide