Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminopyridine-2-carboxylic acid, 98%

Click to view available options
Quantity:
5g
25g
100g
1462-86-8
C6H6N2O2
138.126
MFCD00090153
BOOMHTFCWOJWFO-UHFFFAOYSA-N
3-aminopicolinic acid,3-amino-2-pyridinecarboxylic acid,3-amino-pyridine-2-carboxylic acid,2-pyridinecarboxylic acid, 3-amino,3-amino-2-picolinic acid,3-amino-2-pyridinecarboxylicacid,3-amino-2-carboxypyridine,2-pyridinecarboxylicacid, 3-amino,pubchem2015,3-aminopyridine-2-carboxylic acid, tech.
73836
3-aminopyridine-2-carboxylic acid
C1=CC(=C(N=C1)C(=O)O)N
1462-86-8
C6H6N2O2
138.126
MFCD00090153
BOOMHTFCWOJWFO-UHFFFAOYSA-N
3-aminopicolinic acid,3-amino-2-pyridinecarboxylic acid,3-amino-pyridine-2-carboxylic acid,2-pyridinecarboxylic acid, 3-amino,3-amino-2-picolinic acid,3-amino-2-pyridinecarboxylicacid,3-amino-2-carboxypyridine,2-pyridinecarboxylicacid, 3-amino,pubchem2015,3-aminopyridine-2-carboxylic acid, tech.
73836
3-aminopyridine-2-carboxylic acid
C1=CC(=C(N=C1)C(=O)O)N
Applications
Employed as an important intermediate for syntheses material and pharmaceutical.

Solubility
Soluble in acetone, ethanol and methanol.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Melting Point 218°C to 222°C
Color Brown
Quantity 100g
Beilstein 3600
Formula Weight 138.13
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-Aminopyridine-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : US5175000

storageNote1 : -30° to -10°C