Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminoquinuclidine dihydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6530-09-2
C7H14N2·2ClH
199.12
MFCD00137395
STZHBULOYDCZET-UHFFFAOYNA-N
3-aminoquinuclidine dihydrochloride,1-azabicyclo 2.2.2 octan-3-amine dihydrochloride,quinuclidine, 3-amino-, dihydrochloride,3aminoquinuclidine dihydrochloride,quinuclidin-3-amine dihydrochloride,1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride,1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride,3-amino-1-azabicyclo 2.2.2 octane dihydrochloride,quinuclidine-3-ylamine, chloride, chloride
197853
1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
C1CN2CCC1C(C2)N.Cl.Cl
6530-09-2
C7H14N2·2ClH
199.12
MFCD00137395
STZHBULOYDCZET-UHFFFAOYNA-N
3-aminoquinuclidine dihydrochloride,1-azabicyclo 2.2.2 octan-3-amine dihydrochloride,quinuclidine, 3-amino-, dihydrochloride,3aminoquinuclidine dihydrochloride,quinuclidin-3-amine dihydrochloride,1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride,1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride,3-amino-1-azabicyclo 2.2.2 octane dihydrochloride,quinuclidine-3-ylamine, chloride, chloride
197853
1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
C1CN2CCC1C(C2)N.Cl.Cl
Melting Point 321°C to 323°C
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: soluble
Formula Weight 199.12
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 3-Aminoquinuclidine dihydrochloride, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 229-424-6

RTECSNumber : VD4590000

TSCA : TSCA