Alfa Aesar™ 3-Aminosalicylic acid, 97%

Manufacturer: Alfa Aesar™ B2247877

Quantity 0.1g

 View more versions of this product

Catalog No. AAB2247877

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 570-23-0
    Molecular Formula C7H7NO3
    Formula Weight 153.14
    MDL Number MFCD00010299
    Beilstein 2090427
    Synonym 3-Amino-2-hydroxybenzoic acid
    Chemical Name or Material 3-Aminosalicylic acid

    Specifications

    Formula Weight 153.14
    Quantity 0.1g
    MDL Number MFCD00010299
    Melting Point ≈240°C (dec.)
    Molecular Formula C7H7NO3
    CAS 570-23-0
    Synonym 3-Amino-2-hydroxybenzoic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 2090427
    Chemical Name or Material 3-Aminosalicylic acid
    Assay 97%

    Description

    Applications
    3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.

    Solubility
    Soluble in potassium hydroxide. Slightly soluble in water.

    Notes
    Air and light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit