Alfa Aesar™ 3-Aminothiophenol, 97%

Manufacturer: Alfa Aesar™ B2547322

Quantity 100g

 View more versions of this product

Catalog No. AAB2547322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22948-02-3
    Molecular Formula C6H7NS
    Formula Weight 125.19
    MDL Number MFCD00007791
    UN Number UN3267
    Synonym 3-Aminobenzenethiol; 3-Mercaptoaniline
    Chemical Name or Material 3-Aminothiophenol

    Specifications

    Formula Weight 125.19
    Molecular Formula C6H7NS
    Quantity 100g
    Boiling Point 123° to 125°C (8mmHg)
    MDL Number MFCD00007791
    UN Number UN3267
    Flash Point >100°C (212°F)
    Synonym 3-Aminobenzenethiol; 3-Mercaptoaniline
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
    CAS 22948-02-3
    Chemical Name or Material 3-Aminothiophenol
    Assay 97%

    Description

    Applications
    3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.

    Solubility
    Soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit