Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3-Aminothiophenol, 97%
Click to view available options
Quantity:
5g
25g
100g
Description
Applications
3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Refractive Index | 1.659 |
Density | 1.179 |
Sensitivity | Air sensitive |
Quantity | 25g |
Flash Point | >100°C (212°F) |
Boiling Point | 123°C to 125°C (8mmHg) |
UN Number | UN3267 |
Solubility Information | Soluble in water. |
Formula Weight | 125.19 |
Percent Purity | 97% |
Show More |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H312-H314-H332-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 245-344-4
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen