Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Azabicyclo[3.3.0]octane hydrochloride, 98%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
112626-50-3
C7H14ClN
147.646
MFCD06411019
HVZRRRPCVOJOLJ-UHFFFAOYSA-N
3-azabicyclo 3.3.0 octane hydrochloride,octahydrocyclopenta c pyrrole hydrochloride,3-azabicyclo 3.3.0 octane hcl,3-azabicyclo 3,3,0 octane hcl,3-azabicyclo 3.3.0 octanehydrochloride,1,2,3,3a,4,5,6,6a-octahydrocyclopenta c pyrrole hydrochloride,pubchem9779,cis-octahydro-cyclopenta c pyrrole hydrochloride,c7h14ncl,3-azabicyclo 3.3.0 octane hydrochloride;
21572123
1,2,3,3a,4,5,6,6a-octahydrocyclopenta[c]pyrrole;hydrochloride
C1CC2CNCC2C1.Cl
112626-50-3
C7H14ClN
147.646
MFCD06411019
HVZRRRPCVOJOLJ-UHFFFAOYSA-N
3-azabicyclo 3.3.0 octane hydrochloride,octahydrocyclopenta c pyrrole hydrochloride,3-azabicyclo 3.3.0 octane hcl,3-azabicyclo 3,3,0 octane hcl,3-azabicyclo 3.3.0 octanehydrochloride,1,2,3,3a,4,5,6,6a-octahydrocyclopenta c pyrrole hydrochloride,pubchem9779,cis-octahydro-cyclopenta c pyrrole hydrochloride,c7h14ncl,3-azabicyclo 3.3.0 octane hydrochloride;
21572123
1,2,3,3a,4,5,6,6a-octahydrocyclopenta[c]pyrrole;hydrochloride
C1CC2CNCC2C1.Cl
Quantity 100g
Beilstein 3672489
Formula Weight 147.65
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-Azabicyclo[3.3.0]octane hydrochloride