Learn More
3-Azido-1-propylamine, Thermo Scientific Chemicals
Description
Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.
Solubility
Soluble in DMSO, DMF, DCM, chloroform, THF, and water.Miscible with dimethylsulfoxide, dimethyl formamide, dichloromethane, tetrahydrofuran, chloroform and water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Color | Yellow |
Flash Point | 60°C (140°F) |
Linear Formula | N3CH2CH2CH2NH2 |
Quantity | 100 mg |
UN Number | UN1993 |
Solubility Information | Soluble in DMSO,DMF,DCM,chloroform,THF,and water;Miscible with dimethylsulfoxide,dimethyl formamide,dichloromethane,tetrahydrofuran,chloroform and water. |
Formula Weight | 100.12 |
Physical Form | Liquid |
Chemical Name or Material | 3-Azido-1-propylamine |
Safety and Handling
Hazard Category | H226-H315-H319-H335 |
Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.