Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Benzoyl-1,1,1-trifluoroacetone, 98+%

Click to view available options
Quantity:
25g
100g
500g
326-06-7
C10H7F3O2
216.159
MFCD00000425
VVXLFFIFNVKFBD-UHFFFAOYSA-N
benzoyl-1,1,1-trifluoroacetone,benzoyltrifluoroacetone,4,4,4-trifluoro-1-phenyl-1,3-butanedione,benzoyl trifluoroacetyl methane,3-benzoyl-1,1,1-trifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-phenyl,4,4,4-trifluoro-1-phenyl-butane-1,3-dione,1-benzoyl-3,3,3-trifluoroacetone,.omega.-trifluoroacetyl acetophenone,1-benzoyl-3,3,3-trifluoro-2-propanone
67589
4,4,4-trifluoro-1-phenylbutane-1,3-dione
C1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F
326-06-7
C10H7F3O2
216.159
MFCD00000425
VVXLFFIFNVKFBD-UHFFFAOYSA-N
benzoyl-1,1,1-trifluoroacetone,benzoyltrifluoroacetone,4,4,4-trifluoro-1-phenyl-1,3-butanedione,benzoyl trifluoroacetyl methane,3-benzoyl-1,1,1-trifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-phenyl,4,4,4-trifluoro-1-phenyl-butane-1,3-dione,1-benzoyl-3,3,3-trifluoroacetone,.omega.-trifluoroacetyl acetophenone,1-benzoyl-3,3,3-trifluoro-2-propanone
67589
4,4,4-trifluoro-1-phenylbutane-1,3-dione
C1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F
Applications
3-Benzoyl-1,1,1-trifluoroacetone, is used as a primary and secondary intermediate. It is used as an Intermediate in organic synthesis.

Solubility
Sparingly Soluble in water (0.24 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Flash Point 98°C (208°F)
Density 1.113
Quantity 500g
Boiling Point 224°C
Melting Point 37°C to 40°C
Beilstein 1875083
Solubility Information Sparingly Soluble in water (0.24g/L) (25°C).
Formula Weight 216.16
Percent Purity ≥98%
Chemical Name or Material 3-Benzoyl-1,1,1-trifluoroacetone

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-307-8

TSCA : Yes

Recommended Storage : Ambient temperatures