Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzoylbenzoic acid, 99%, Acros Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
579-18-0
C14H10O3
226.23
AXJXRLHTQQONQR-UHFFFAOYSA-N
benzoic acid, 3-benzoyl,m-benzoylbenzoic acid,3-benzoyl benzoic acid,3-benzoyl-benzoic acid,3-phenylcarbonyl benzoic acid,3-benzoylbenzoicacid,3-benzoyl-benzoesaure,3-benzoylbenzoic-acid,acmc-20an9k,benzoic acid,3-benzoyl
101386
3-benzoylbenzoic acid
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)C(=O)O
579-18-0
C14H10O3
226.23
AXJXRLHTQQONQR-UHFFFAOYSA-N
benzoic acid, 3-benzoyl,m-benzoylbenzoic acid,3-benzoyl benzoic acid,3-benzoyl-benzoic acid,3-phenylcarbonyl benzoic acid,3-benzoylbenzoicacid,3-benzoyl-benzoesaure,3-benzoylbenzoic-acid,acmc-20an9k,benzoic acid,3-benzoyl
101386
3-benzoylbenzoic acid
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)C(=O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color White to Beige
Melting Point 161.0°C to 166.5°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Linear Formula C6H5COC6H4COOH
Beilstein 10,752
Formula Weight 226.23
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 3-Benzoylbenzoic acid, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

TSCA : TSCA