Alfa Aesar™ 3-[Benzyl(ethyl)carbamoyl]-4-fluorobenzeneboronic acid, 97%

Manufacturer: Alfa Aesar™ H54914MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH54914MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C16H17BFNO3
    Formula Weight 301.12
    MDL Number MFCD20040268
    Chemical Name or Material 3-[Benzyl(ethyl)carbamoyl]-4-fluorobenzeneboronic acid

    Specifications

    Formula Weight 301.12
    Molecular Formula C16H17BFNO3
    MDL Number MFCD20040268
    Quantity 250mg
    Chemical Name or Material 3-[Benzyl(ethyl)carbamoyl]-4-fluorobenzeneboronic acid
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit